Assay Details
| Gene Symbol: acsl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003569.2 | NP_001003569.1 | 2 - 3 | 316 | 82 | |
| XM_005169565.4 | XP_005169622.1 | 2 - 3 | 366 | 82 | ||
| GenBank mRNA | BC077120.1 | - | 2 - 3 | 317 | 82 | |
More Information
Set Membership: |
|

acsl1b
Genomic Map