Assay Details
| Gene Symbol: mfsd2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003570.2 | NP_001003570.1 | 8 - 9 | 1130 | 69 | |
| GenBank mRNA | BC077117.1 | - | 8 - 9 | 1077 | 69 | |
More Information
Set Membership: |
|

mfsd2ab
Genomic Map