Assay Details
| Gene Symbol: epb41l3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003987.1 | NP_001003987.1 | 18 - 19 | 2259 | 65 | |
| XM_021470150.1 | XP_021325825.1 | 20 - 21 | 2923 | 65 | ||
| XM_021470151.1 | XP_021325826.1 | 20 - 21 | 2922 | 65 | ||
| XM_021470153.1 | XP_021325828.1 | 18 - 19 | 2318 | 65 | ||
| XM_021470154.1 | XP_021325829.1 | 18 - 19 | 2265 | 65 | ||
| GenBank mRNA | BC080246.1 | - | 18 - 19 | 2259 | 65 | |
| BC158197.1 | - | 18 - 19 | 2265 | 65 | ||
More Information
Set Membership: |
|

epb41l3a
Genomic Map