Assay Details
| Gene Symbol: epb41l3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003987.1 | NP_001003987.1 | 14 - 15 | 1730 | 72 | |
| XM_021470142.1 | XP_021325817.1 | 15 - 16 | 1842 | 72 | ||
| XM_021470143.1 | XP_021325818.1 | 14 - 15 | 1796 | 72 | ||
| XM_021470144.1 | XP_021325819.1 | 15 - 16 | 1841 | 72 | ||
| XM_021470145.1 | XP_021325820.1 | 15 - 16 | 1834 | 72 | ||
| XM_021470146.1 | XP_021325821.1 | 15 - 16 | 2014 | 72 | ||
| XM_021470147.1 | XP_021325822.1 | 15 - 16 | 1873 | 72 | ||
| XM_021470148.1 | XP_021325823.1 | 16 - 17 | 1954 | 72 | ||
| XM_021470149.1 | XP_021325824.1 | 15 - 16 | 1791 | 72 | ||
| XM_021470150.1 | XP_021325825.1 | 15 - 16 | 1839 | 72 | ||
| XM_021470151.1 | XP_021325826.1 | 15 - 16 | 1838 | 72 | ||
| XM_021470152.1 | XP_021325827.1 | 15 - 16 | 1840 | 72 | ||
| XM_021470153.1 | XP_021325828.1 | 14 - 15 | 1789 | 72 | ||
| XM_021470154.1 | XP_021325829.1 | 14 - 15 | 1736 | 72 | ||
| XM_021470155.1 | XP_021325830.1 | 4 - 5 | 461 | 72 | ||
| GenBank mRNA | BC080246.1 | - | 14 - 15 | 1730 | 72 | |
| BC158197.1 | - | 14 - 15 | 1736 | 72 | ||
More Information
Set Membership: |
|

epb41l3a
Genomic Map