Assay Details
| Gene Symbol: epb41l3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003987.1 | NP_001003987.1 | 16 - 17 | 2024 | 68 | |
| XM_021470152.1 | XP_021325827.1 | 17 - 18 | 2134 | 68 | ||
| XM_021470153.1 | XP_021325828.1 | 16 - 17 | 2083 | 68 | ||
| XM_021470154.1 | XP_021325829.1 | 16 - 17 | 2030 | 68 | ||
| GenBank mRNA | BC080246.1 | - | 16 - 17 | 2024 | 68 | |
| BC158197.1 | - | 16 - 17 | 2030 | 68 | ||
More Information
Set Membership: |
|

epb41l3a
Genomic Map