Assay Details
| Gene Symbol: elf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004116.1 | NP_001004116.1 | 4 - 5 | 701 | 89 | |
| XM_005169610.4 | XP_005169667.1 | 4 - 5 | 742 | 89 | ||
| XM_009291253.3 | XP_009289528.1 | 2 - 3 | 164 | 89 | ||
| GenBank mRNA | BC080260.1 | - | 4 - 5 | 701 | 89 | |
More Information
Set Membership: |
|

elf2a
Genomic Map