Assay Details
| Gene Symbol: clpxa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004581.1 | NP_001004581.1 | 6 - 7 | 1055 | 79 | |
| XM_021477471.1 | XP_021333146.1 | 7 - 8 | 79 | |||
| GenBank mRNA | BC081643.1 | - | 6 - 7 | 1055 | 79 | |
More Information
Set Membership: |
|

clpxa
Genomic Map