Assay Details
| Gene Symbol: fkbp1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005594.1 | NP_001005594.1 | 2 - 3 | 126 | 87 | |
| GenBank mRNA | BC081522.1 | - | 2 - 3 | 126 | 87 | |
More Information
Set Membership: |
|

fkbp1ab
Genomic Map