Assay Details
| Gene Symbol: chchd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006051.1 | NP_001006051.1 | 2 - 3 | 312 | 97 | |
| GenBank mRNA | BC083285.1 | - | 2 - 3 | 312 | 97 | |
| BC164901.1 | - | 2 - 3 | 159 | 97 | ||
More Information
Set Membership: |
|

chchd4b
Genomic Map