Assay Details
| Gene Symbol: scp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006093.2 | NP_001006093.1 | 4 - 5 | 454 | 76 | |
| GenBank mRNA | BC083225.1 | - | 4 - 5 | 409 | 76 | |
| BC155252.1 | - | 4 - 5 | 411 | 76 | ||
More Information
Set Membership: |
|

scp2b
Genomic Map