Assay Details
| Gene Symbol: rnf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007290.2 | NP_001007291.2 | 4 - 5 | 564 | 87 | |
| GenBank mRNA | AY621088.2 | - | 4 - 5 | 499 | 87 | |
| BC071534.1 | - | 4 - 5 | 87 | |||
More Information
Set Membership: |
|

rnf141
Genomic Map