Assay Details
| Gene Symbol: dennd2db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007346.1 | NP_001007347.1 | 3 - 4 | 359 | 56 | |
| GenBank mRNA | BC085532.1 | - | 3 - 4 | 359 | 56 | |
More Information
Set Membership: |
|

dennd2db
Genomic Map