Assay Details
| Gene Symbol: fgf18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012379.1 | NP_001012379.1 | 2 - 3 | 77 | 81 | |
| GenBank mRNA | AB195468.1 | - | 2 - 3 | 77 | 81 | |
| BC163126.1 | - | 2 - 3 | 162 | 81 | ||
| BC163136.1 | - | 2 - 3 | 159 | 81 | ||
More Information
Set Membership: |
|

fgf18b
Genomic Map