Assay Details
| Gene Symbol: ptbp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328429.1 | NP_001315358.1 | 10 - 11 | 1138 | 95 | |
| XM_017353829.2 | XP_017209318.1 | 10 - 11 | 1081 | 95 | ||
| XM_017353831.2 | XP_017209320.1 | 5 - 6 | 617 | 95 | ||
| GenBank mRNA | BC090482.1 | - | 4 - 5 | 291 | 95 | |
| BC165254.1 | - | 1 - 2 | 68 | 95 | ||
More Information
Set Membership: |
|

ptbp2a
Genomic Map