Assay Details
| Gene Symbol: ptbp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328429.1 | NP_001315358.1 | 13 - 14 | 1532 | 89 | |
| XM_017353829.2 | XP_017209318.1 | 13 - 14 | 1475 | 89 | ||
| XM_017353831.2 | XP_017209320.1 | 8 - 9 | 1011 | 89 | ||
| GenBank mRNA | BC090482.1 | - | 7 - 8 | 685 | 89 | |
| BC165254.1 | - | 4 - 5 | 462 | 89 | ||
More Information
Set Membership: |
|

ptbp2a
Genomic Map