Assay Details
| Gene Symbol: ablim1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009306617.3 | XP_009304892.1 | 8 - 9 | 1195 | 78 | |
| XM_009306619.3 | XP_009304894.1 | 8 - 9 | 1194 | 78 | ||
| XM_009306622.3 | XP_009304897.1 | 8 - 9 | 1193 | 78 | ||
| XM_009306623.3 | XP_009304898.1 | 8 - 9 | 1192 | 78 | ||
| XM_009306624.3 | XP_009304899.1 | 8 - 9 | 1194 | 78 | ||
| XM_009306625.3 | XP_009304900.1 | 3 - 4 | 299 | 78 | ||
| XM_009306627.3 | XP_009304902.1 | 8 - 9 | 1194 | 78 | ||
| XM_017358455.2 | XP_017213944.1 | 8 - 9 | 1195 | 78 | ||
| XM_017358456.2 | XP_017213945.1 | 8 - 9 | 1193 | 78 | ||
| XM_021480098.1 | XP_021335773.1 | 8 - 9 | 1194 | 78 | ||
| GenBank mRNA | BC091932.1 | - | 8 - 9 | 1026 | 78 | |
| EB912805.1 | - | 3 - 4 | 250 | 78 | ||
More Information
Set Membership: |
|

ablim1b
Genomic Map