Assay Details
| Gene Symbol: erc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015065.1 | NP_001015065.1 | 10 - 11 | 2248 | 107 | |
| XM_009298006.3 | XP_009296281.1 | 11 - 12 | 2592 | 107 | ||
| XM_021470487.1 | XP_021326162.1 | 10 - 11 | 2242 | 107 | ||
| GenBank mRNA | AY916778.1 | - | 10 - 11 | 2248 | 107 | |
More Information
Set Membership: |
|

erc1a
Genomic Map