Assay Details
| Gene Symbol: erc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015065.1 | NP_001015065.1 | 1 - 2 | 692 | 120 | |
| XM_009298003.3 | XP_009296278.1 | 2 - 3 | 1037 | 120 | ||
| XM_009298005.3 | XP_009296280.1 | 2 - 3 | 1038 | 120 | ||
| XM_009298006.3 | XP_009296281.1 | 2 - 3 | 1036 | 120 | ||
| XM_021470486.1 | XP_021326161.1 | 1 - 2 | 767 | 120 | ||
| XM_021470487.1 | XP_021326162.1 | 1 - 2 | 686 | 120 | ||
| GenBank mRNA | AY916778.1 | - | 1 - 2 | 692 | 120 | |
| BC171689.1 | - | 1 - 2 | 767 | 120 | ||
More Information
Set Membership: |
|

erc1a
Genomic Map