Assay Details
| Gene Symbol: erc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015065.1 | NP_001015065.1 | 4 - 5 | 1303 | 104 | |
| XM_009298003.3 | XP_009296278.1 | 5 - 6 | 1648 | 104 | ||
| XM_009298005.3 | XP_009296280.1 | 5 - 6 | 1649 | 104 | ||
| XM_009298006.3 | XP_009296281.1 | 5 - 6 | 1647 | 104 | ||
| XM_021470486.1 | XP_021326161.1 | 4 - 5 | 1378 | 104 | ||
| XM_021470487.1 | XP_021326162.1 | 4 - 5 | 1297 | 104 | ||
| GenBank mRNA | AY916778.1 | - | 4 - 5 | 1303 | 104 | |
| BC171689.1 | - | 4 - 5 | 1378 | 104 | ||
More Information
Set Membership: |
|

erc1a
Genomic Map