Assay Details
| Gene Symbol: erc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015065.1 | NP_001015065.1 | 8 - 9 | 1918 | 134 | |
| XM_009298003.3 | XP_009296278.1 | 9 - 10 | 2263 | 134 | ||
| XM_009298005.3 | XP_009296280.1 | 9 - 10 | 2264 | 134 | ||
| XM_009298006.3 | XP_009296281.1 | 9 - 10 | 2262 | 134 | ||
| XM_021470486.1 | XP_021326161.1 | 8 - 9 | 1993 | 134 | ||
| XM_021470487.1 | XP_021326162.1 | 8 - 9 | 1912 | 134 | ||
| GenBank mRNA | AY916778.1 | - | 8 - 9 | 1918 | 134 | |
| BC171689.1 | - | 8 - 9 | 1993 | 134 | ||
More Information
Set Membership: |
|

erc1a
Genomic Map