Assay Details
| Gene Symbol: gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017560.1 | NP_001017560.1 | 6 - 7 | 843 | 62 | |
| XM_005167557.4 | XP_005167614.1 | 6 - 7 | 1549 | 62 | ||
| XM_017357671.2 | XP_017213160.1 | 5 - 6 | 869 | 62 | ||
| XM_021478649.1 | XP_021334324.1 | 5 - 6 | 872 | 62 | ||
| GenBank mRNA | BC092932.1 | - | 6 - 7 | 843 | 62 | |
More Information
Set Membership: |
|

gtdc1
Genomic Map