Assay Details
| Gene Symbol: mrpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017696.1 | NP_001017696.1 | 4 - 5 | 602 | 76 | |
| XM_009299749.3 | XP_009298024.1 | 5 - 6 | 637 | 76 | ||
| GenBank mRNA | BC093312.1 | - | 4 - 5 | 602 | 76 | |
More Information
Set Membership: |
|

mrpl12
Genomic Map