Assay Details
| Gene Symbol: ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024207.2 | NP_001019378.1 | 7 - 8 | 903 | 66 | |
| GenBank mRNA | BC095838.1 | - | 7 - 8 | 905 | 66 | |
| BC165879.1 | - | 7 - 8 | 856 | 66 | ||
More Information
Set Membership: |
|

ugcg
Genomic Map