Assay Details
| Gene Symbol: csnk1g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024379.1 | NP_001019550.1 | 5 - 6 | 1003 | 112 | |
| XM_005171206.4 | XP_005171263.1 | 6 - 7 | 1405 | 112 | ||
| XM_005171207.4 | XP_005171264.1 | 6 - 7 | 1405 | 112 | ||
| GenBank mRNA | BC095384.1 | - | 5 - 6 | 1003 | 112 | |
More Information
Set Membership: |
|

csnk1g2a
Genomic Map