Assay Details
| Gene Symbol: derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024395.1 | NP_001019566.1 | 1 - 2 | 105 | 77 | |
| GenBank mRNA | BC095262.1 | - | 1 - 2 | 105 | 77 | |
| BC165145.1 | - | 1 - 2 | 134 | 77 | ||
More Information
Set Membership: |
|

derl2
Genomic Map