Assay Details
| Gene Symbol: ptgs2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025504.2 | NP_001020675.1 | 3 - 4 | 358 | 98 | |
| GenBank mRNA | BC139568.1 | - | 3 - 4 | 375 | 98 | |
| DQ494791.1 | - | 3 - 4 | 358 | 98 | ||
More Information
Set Membership: |
|

ptgs2b
Genomic Map