Assay Details
| Gene Symbol: ptgs2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025504.2 | NP_001020675.1 | 5 - 6 | 682 | 91 | |
| GenBank mRNA | BC139568.1 | - | 5 - 6 | 699 | 91 | |
| DQ494791.1 | - | 5 - 6 | 682 | 91 | ||
More Information
Set Membership: |
|

ptgs2b
Genomic Map