Assay Details
| Gene Symbol: ptgs2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025504.2 | NP_001020675.1 | 7 - 8 | 1015 | 86 | |
| GenBank mRNA | DQ494791.1 | - | 7 - 8 | 1015 | 86 | |
More Information
Set Membership: |
|

ptgs2b
Genomic Map