Assay Details
| Gene Symbol: ano10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030206.1 | NP_001025377.1 | 8 - 9 | 1342 | 87 | |
| GenBank mRNA | BC097195.1 | - | 8 - 9 | 1342 | 87 | |
More Information
Set Membership: |
|

ano10a
Genomic Map