Assay Details
| Gene Symbol: nr6a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033720.1 | NP_001028892.1 | 5 - 6 | 620 | 138 | |
| GenBank mRNA | BC097237.1 | - | 5 - 6 | 620 | 138 | |
| DQ017610.1 | - | 3 - 4 | 433 | 138 | ||
More Information
Set Membership: |
|

nr6a1b
Genomic Map