Assay Details
| Gene Symbol: cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033734.1 | NP_001028906.1 | 2 - 3 | 405 | 105 | |
| GenBank mRNA | BC097138.1 | - | 2 - 3 | 405 | 105 | |
More Information
Set Membership: |
|

cep19
Genomic Map