Assay Details
| Gene Symbol: pi4kaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034972.1 | NP_001030144.1 | 35 - 36 | 4109 | 85 | |
| XM_005172596.4 | XP_005172653.1 | 36 - 37 | 4337 | 85 | ||
| GenBank mRNA | AY929292.1 | - | 35 - 36 | 4109 | 85 | |
More Information
Set Membership: |
|

pi4kaa
Genomic Map