Assay Details
| Gene Symbol: pi4kaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034972.1 | NP_001030144.1 | 3 - 4 | 286 | 79 | |
| XM_005172596.4 | XP_005172653.1 | 4 - 5 | 514 | 79 | ||
| XM_009305081.3 | XP_009303356.1 | 4 - 5 | 515 | 79 | ||
| GenBank mRNA | AY929292.1 | - | 3 - 4 | 286 | 79 | |
More Information
Set Membership: |
|

pi4kaa
Genomic Map