Assay Details
| Gene Symbol: zyg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040332.2 | NP_001035422.1 | 7 - 8 | 1663 | 68 | |
| GenBank mRNA | BC115195.1 | - | 7 - 8 | 1629 | 68 | |
More Information
Set Membership: |
|

zyg11
Genomic Map