Assay Details
| Gene Symbol: tnfrsf9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040369.1 | NP_001035459.1 | 3 - 4 | 305 | 86 | |
| XM_009296991.3 | XP_009295266.1 | 3 - 4 | 813 | 86 | ||
| XM_009296992.3 | XP_009295267.1 | 3 - 4 | 817 | 86 | ||
| GenBank mRNA | BC115175.1 | - | 3 - 4 | 305 | 86 | |
More Information
Set Membership: |
|

tnfrsf9a
Genomic Map