Assay Details
| Gene Symbol: tnfrsf9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040369.1 | NP_001035459.1 | 6 - 7 | 601 | 145 | |
| GenBank mRNA | BC115175.1 | - | 6 - 7 | 601 | 145 | |
More Information
Set Membership: |
|

tnfrsf9a
Genomic Map