Assay Details
| Gene Symbol: klf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044766.2 | NP_001038231.1 | 1 - 2 | 454 | 80 | |
| GenBank mRNA | BC124329.1 | - | 1 - 2 | 454 | 80 | |
| BC150423.1 | - | 1 - 2 | 453 | 80 | ||
| BC165378.1 | - | 1 - 2 | 132 | 80 | ||
More Information
Set Membership: |
|

klf7b
Genomic Map