Assay Details
| Gene Symbol: klf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044766.2 | NP_001038231.1 | 3 - 4 | 1188 | 61 | |
| GenBank mRNA | BC124329.1 | - | 3 - 4 | 1188 | 61 | |
| BC165378.1 | - | 3 - 4 | 866 | 61 | ||
More Information
Set Membership: |
|

klf7b
Genomic Map