Assay Details
| Gene Symbol: kcnh5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276280.1 | NP_001263209.1 | 4 - 5 | 501 | 63 | |
| GenBank mRNA | BC163622.1 | - | 4 - 5 | 501 | 63 | |
More Information
Set Membership: |
|

kcnh5a
Genomic Map