Assay Details
| Gene Symbol: ppp2r5cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044821.3 | NP_001038286.2 | 1563 | 72 | ||
| NM_001362603.1 | NP_001349532.1 | 1413 | 72 | |||
| XM_005160898.4 | XP_005160955.1 | 14 - 15 | 1564 | 72 | ||
| GenBank mRNA | BC139635.1 | - | 14 - 15 | 1536 | 72 | |
More Information
Set Membership: |
|

ppp2r5cb
Genomic Map