Assay Details
| Gene Symbol: slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044846.1 | NP_001038311.1 | 3 - 4 | 978 | 80 | |
| XM_009294668.3 | XP_009292943.1 | 3 - 4 | 1033 | 80 | ||
| XM_017352483.2 | XP_017207972.1 | 3 - 4 | 1032 | 80 | ||
| XM_017352484.2 | XP_017207973.1 | 3 - 4 | 1031 | 80 | ||
| GenBank mRNA | BC125960.1 | - | 3 - 4 | 462 | 80 | |
| BC133831.1 | - | 3 - 4 | 987 | 80 | ||
More Information
Set Membership: |
|

slain2
Genomic Map