Assay Details
| Gene Symbol: dvl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044917.1 | NP_001038382.1 | 10 - 11 | 1038 | 85 | |
| XM_005169297.4 | XP_005169354.1 | 10 - 11 | 1456 | 85 | ||
| XM_005169298.4 | XP_005169355.1 | 10 - 11 | 1456 | 85 | ||
| XM_005169299.4 | XP_005169356.1 | 8 - 9 | 1274 | 85 | ||
| XM_005169301.4 | XP_005169358.1 | 10 - 11 | 1456 | 85 | ||
| GenBank mRNA | BC171461.1 | - | 10 - 11 | 1147 | 85 | |
More Information
Set Membership: |
|

dvl1b
Genomic Map