Assay Details
| Gene Symbol: dvl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044917.1 | NP_001038382.1 | 11 - 12 | 1110 | 78 | |
| XM_005169298.4 | XP_005169355.1 | 11 - 12 | 1528 | 78 | ||
| GenBank mRNA | BC171461.1 | - | 11 - 12 | 1219 | 78 | |
More Information
Set Membership: |
|

dvl1b
Genomic Map