Assay Details
| Gene Symbol: dvl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044917.1 | NP_001038382.1 | 13 - 14 | 1412 | 114 | |
| XM_005169297.4 | XP_005169354.1 | 12 - 13 | 1908 | 114 | ||
| XM_005169298.4 | XP_005169355.1 | 13 - 14 | 1830 | 114 | ||
| XM_005169299.4 | XP_005169356.1 | 10 - 11 | 1726 | 114 | ||
| XM_005169301.4 | XP_005169358.1 | 12 - 13 | 1908 | 114 | ||
More Information
Set Membership: |
|

dvl1b
Genomic Map