Assay Details
| Gene Symbol: npr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044937.2 | NP_001038402.1 | 17 - 18 | 2715 | 83 | |
| XM_005170050.4 | XP_005170107.1 | 18 - 19 | 3395 | 83 | ||
| GenBank mRNA | GU076492.1 | - | 2 - 3 | 254 | 83 | |
More Information
Set Membership: |
|

npr1a
Genomic Map