Assay Details
| Gene Symbol: npr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044937.2 | NP_001038402.1 | 9 - 10 | 1703 | 72 | |
| XM_005170050.4 | XP_005170107.1 | 10 - 11 | 2383 | 72 | ||
More Information
Set Membership: |
|

npr1a
Genomic Map