Assay Details
| Gene Symbol: mfsd12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044972.1 | NP_001038437.1 | 8 - 9 | 1422 | 96 | |
| XM_005171209.4 | XP_005171266.1 | 8 - 9 | 1774 | 96 | ||
| GenBank mRNA | BC128826.1 | - | 8 - 9 | 1423 | 96 | |
More Information
Set Membership: |
|

mfsd12a
Genomic Map