Assay Details
| Gene Symbol: p4ha1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044998.1 | NP_001038463.1 | 7 - 8 | 982 | 125 | |
| XM_009306501.3 | XP_009304776.1 | 8 - 9 | 1251 | 125 | ||
More Information
Set Membership: |
|

p4ha1a
Genomic Map