Assay Details
| Gene Symbol: p4ha1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044998.1 | NP_001038463.1 | 9 - 10 | 1116 | 61 | |
| XM_009306502.3 | XP_009304777.1 | 9 - 10 | 1319 | 61 | ||
More Information
Set Membership: |
|

p4ha1a
Genomic Map