Assay Details
| Gene Symbol: gna11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045036.2 | NP_001038501.1 | 4 - 5 | 596 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gna11a| Gene Symbol: gna11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045036.2 | NP_001038501.1 | 4 - 5 | 596 | 85 | |
Set Membership: |
|